Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 10.0 N, from NF/USP material, Ricca Chemical
SDP

Catalog No. R7470050120 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R7470050120 120 mL
R74700501A 1 L
R74700504A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R7470050120 Supplier Ricca Chemical Company Supplier No. R7470050120A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2

Specifications

Color Colorless
CAS Max % 69.32, 30.68
Molecular Formula H2O
Concentration 9.95 to 10.05 N
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Quantity 120 mL
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Assay Normal at 20°C
Chemical Name or Material Sodium Hydroxide Solution
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.