Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 10% (w/v) (100 g/L), Ricca Chemical
SDP

Catalog No. 72605 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
72605 20 L Cubitainer
7260-4 120 mL Natural Poly Bottle
7260-16 500 mL Natural Poly Bottle
7260-32 1 L Natural Poly Bottle
7260-1 4 L Natural Poly Bottle
72602-5 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 72605 Supplier Ricca Chemical Company Supplier No. 72605
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Quantity 20 L
Percent Purity 9.96 to 10.04%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (w/v) Aqueous Solution (100g/L)
Packaging Cubitainer
Chemical Name or Material Sodium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.