Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium hydroxide solution, 1310-73-2, MFCD00003548, 100 mL
Catalog No. NC0310325
Encompass
Linear Formula: NaOH, Molecular Weight: 40.00, BioUltra, for molecular biology, 10 M in H2O.
Catalog No. NC0310325 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 72068100ML
Linear Formula: NaOH, Molecular Weight: 40.00, BioUltra, for molecular biology, 10 M in H2O.