Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium Hydroxide, 1310-73-2, MFCD00003548, 500G
Catalog No. NC1385199
Encompass
Molecular Formula: NaOH, Molecular Weight: 40.00, Assay: ≥97%, Solubility: water: soluble 1260 g/L at 20 °C
Catalog No. NC1385199 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 795429500G
Molecular Formula: NaOH, Molecular Weight: 40.00, Assay: ≥97%, Solubility: water: soluble 1260 g/L at 20 °C