Learn More
Chemical Services Laboratories Sodium hydroxide, 1310-73-2, MFCD00003548, Box of 12 ampules
Catalog No. NC9147577
Encompass
Molecular Formula NaOH, Molecular Weight 40.00; Sodium Hydroxide for standardizing 0.10 N Acid
Catalog No. NC9147577 Supplier Chemical Services Laboratories Supplier No. 103
Molecular Formula NaOH, Molecular Weight 40.00; Sodium Hydroxide for standardizing 0.10 N Acid