Learn More
Electron Microscopy Sciences Sodium Hydroxide 1N Solution 225 ML
SDP
Catalog No. 5028741
Encompass_Preferred
Carbonate-free. This solution is prepared from ACS Reagent grade sodium hydroxide, and is standardized colormetrically against potassium biphthalate primary reference material. Normal Concentration 1.98-2.02
This item is not returnable. View return policy
Catalog No. 50-287-41 Supplier Electron Microscopy Sciences Supplier No. 2117001
Carbonate-free. This solution is prepared from ACS Reagent grade sodium hydroxide, and is standardized colormetrically against potassium biphthalate primary reference material. Normal Concentration 1.98-2.02