Learn More
Aqua Solutions SODIUM HYDROXIDE 2.0% 1L
SDP
Catalog No. NC2070190
Encompass_Preferred
Sodium hydroxide solution, 8, UN1824, PG III,Size: 1L. Actual Assay w/w = 1.9900 – 2.0100
Catalog No. NC2070190 Supplier Aqua Solutions Supplier No. 86311L
Sodium hydroxide solution, 8, UN1824, PG III,Size: 1L. Actual Assay w/w = 1.9900 – 2.0100