Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 2.50 N, Ricca Chemical
SDP

Catalog No. 745716 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
25 mL HDPE Bottle
Cubitainer
Natural Poly Bottle
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
745716 500 mL Natural Poly Bottle
01-213-128 25 mL 25 mL HDPE Bottle
7457-32 1 L Natural Poly Bottle
7457-1 4 L Natural Poly Bottle
R745700010F 10 L Cubitainer
7457-5 20 L Cubitainer
745755 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 745716 Supplier Ricca Chemical Company Supplier No. 745716
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1310-73-2 | HNaO | 40.00 g/mol

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 89.0, 9.08
CAS Max % 93.0, 9.45
Concentration or Composition Notes 2.50Normal
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Quantity 500 mL
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.