Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 2% (w/v), Ricca Chemical
SDP

Catalog No. 01213118 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-213-118 1 L
01-213-113 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-213-118 Supplier Ricca Chemical Company Supplier No. R72442001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Sodium Hydroxide
Color Colorless
CAS 1310-73-2, 7732-18-5
Boiling Point 100°C
Melting Point 0.0°C
CAS Max % 1.96%, 98.04%
CAS Min % 98.04%, 1.96%
Quantity 1 L
Percent Purity 2%
Grade Laboratory
Packaging 1 L HDPE Bottle
Specific Gravity 2% (w/v)
Traceability to NIST NIST SRM# 723
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.