Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 20% w/v P4TF8-S12 Class F, Reagents

Catalog No. 01333885
Change view
Click to view available options
Quantity:
500 mL
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-885 10 L
01-333-880 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-885 Supplier Reagents Holdings Llc Supplier No. CS10100010F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Hydroxide, 20% w/v P4TF8-S12 Class F, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH >13
Color Colorless
Boiling Point 105°C
Quantity 10 L
Percent Purity 20%
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.1873
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 20% w/v P4TF8-S12 Class F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.