Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 20% (w/w), Ricca Chemical
SDP

Catalog No. 72715HP Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
72715HP 20 L Hard Pack
72-711 4 L Natural Poly Bottle
72715 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 72715HP Supplier Ricca Chemical Company Supplier No. 72715HP
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 66.0, 32.16
CAS Max % 69.0, 33.48
Percent Purity 20%
Grade Laboratory
Packaging Hard Pack
Solvent or Matrix Aqueous Solution
pH >13
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 20% (w/w)
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.