Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 3.0 Normal Standardized, Reagents

Catalog No. 01333734
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-734 1 L
01-333-733 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-734 Supplier Reagents Holdings Llc Supplier No. CS1303001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Hydroxide, 3.0 Normal Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Hydroxide, 3.0 Normal Standardized
pH 14
Color Colorless
Boiling Point 100°C
CAS 7732-18-5, 1310-73-2
Molecular Formula NaOH/H2O
Synonym Lye, Caustic soda
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1192
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.