Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 4.00 N, Ricca Chemical
SDP

Catalog No. R74600004F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R74600004F 4 L Cubitainer
7460-32 1 L Natural Poly Bottle
7460-1 4 L Natural Poly Bottle
7460-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R74600004F Supplier Ricca Chemical Company Supplier No. 74601CT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1310-73-2 | 40.00 g/mol

Chemical Identifiers

CAS 1310-73-2

Specifications

Quantity 4 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 4.00N
Packaging Cubitainer
Chemical Name or Material Sodium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.