Learn More
Sodium Hydroxide, 40% (w/w), Analytical Reagent Grade, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Quantity | 4 L |
| IUPAC Name | sodium hydroxide |
| Molecular Weight (g/mol) | 40.00 |
| Percent Purity | 39.8 to 40.2% |
| Grade | Analytical |
| Concentration or Composition (by Analyte or Components) | 40% (w/w) Aqueous Solution |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Sodium Hydroxide, Nitrogen Free |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.