Learn More
Aqua Solutions Sodium Hydroxide 5% w/w Certified Solution (250ML)
SDP
Catalog No. NC9195178
Encompass_Preferred
Sodium Hydroxide 5% w/w Certified Solution (250ML)
Catalog No. NC9195178 Supplier Aqua Solutions Supplier No. SYN015
Sodium Hydroxide 5% w/w Certified Solution (250ML)