Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 5 N Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18606552 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
4 x 4 L
6 x 1 L
55 gal.
Packaging:
Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-552 1 L Poly Bottle
18-606-553 4 L Poly Bottle
18-606-558 6 x 1 L Poly Bottle
18-606-554 4 x 4 L Poly Bottle
18-606-555 20 L Poly Bottle
18-606-557 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-606-552 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S3901LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S-390, 1310-73-2

Spectrum™ Chemical Sodium Hydroxide, 5 N Aqueous Solution is a standard solution of Caustic soda ideally suited for laboratory titrimetric techniques. You can rely on the quality of LabChem analytical test solutions for lot-to-lot reproducibility.

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.17, 0.83
Concentration 5.0 N ± 0.2 N
Quantity 1 L
Grade Chemical Solution
Packaging Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.