Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 50% (w/w) (760 g/L), Purified Grade, Ricca Chemical
SDP

Catalog No. R729000055E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
15 g
500 mL
1 L
20 L
55 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R729000055E 55 gal. Poly Drum
729016 500 mL Natural Poly Bottle
729015G 15 g Poly Drum
729032 1 L Natural Poly Bottle
7290-5 20 L Cubitainer
7290-5HP 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R729000055E Supplier Ricca Chemical Company Supplier No. R729000055E
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrogen Free - Suitable for Kjeldahl Nitrogen Analysis

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 49.0
CAS Max % 51.0
Synonym sodium hydroxide
Quantity 55 gal.
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Percent Purity min 49.5% w/w
Grade Purified
Packaging Poly Drum
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.