Learn More
Sodium hydroxide, 50% w/w aq. soln., Thermo Scientific Chemicals
Description
Sodium hydroxide, 50% w/w aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 5 kg |
Solubility Information | Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether. |
Formula Weight | 40 |
Percent Purity | 50% |
Appearance | Colorless liquid |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Density | 1.53 |
Chemical Name or Material | Sodium hydroxide |
Safety and Handling
Hazard Category | H290-H314-H335 |
Precautionary Statement | P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
DOTInformation | DOT Class: 8; Packing Group: II |
EINECSNumber | 215-185-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.