Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, 50% w/w aq. soln.

Catalog No. AA33382A1
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33382A1 1 kg
AA3338222 100 g
AA33382A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA33382A1 Supplier Thermo Scientific Chemicals Supplier No. 033382A1
Add to Cart

CAS: 1310-73-2 | HNaO | 39.997 g/mol

Sodium hydroxide, 50% w/w aq. solution is used as a chemical reactant or neutralization agent in various industries like chemical, pharmaceutical, textile, pulp and paper, metal and food industries and soap and detergent manufacturing. It is also used in the manufacture of synthetic fabrics such as rayon and also for manufacturing food products. It is utilized for pH adjustment and formation of in situ sodium methylate in bioethanol and biodiesel processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 kg
Solubility Information Miscible with glycerol,ethanol and methanol. Immiscible with acetone and ether.
Formula Weight 40
Percent Purity 50%
Appearance Colorless liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Density 1.53 g/mL
Chemical Name or Material Sodium hydroxide
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation DOT Class: 8; Packing Group: II
EINECSNumber 215-185-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.