Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 50% (w/w) (760 g/L), Analytical Reagent Grade, Ricca Chemical
SDP

Catalog No. R729100020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
30 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R729100020F 20 L Cubitainer
7291-16 500 mL Natural Poly Bottle
729132 1 L Natural Poly Bottle
7291-1 4 L Natural Poly Bottle
72915HP 20 L Hard Pack
729130 30 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R729100020F Supplier Ricca Chemical Company Supplier No. R729100020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES [OH-].[Na+]

Specifications

pH >13
Color Colorless
Boiling Point 100°C
Melting Point 12°C
Solubility Information Miscible
Quantity 20 L
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40
Percent Purity 50 to 52% w/w
Grade Analytical
Packaging Cubitainer
Vapor Pressure 13 mmHg at 60°C
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.