Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 50% (w/w), Solution, 48-52%, Spectrum™ Chemical
SDP

Catalog No. 18613615 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
2.5 gal.
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-615 100 mL Poly Bottle
18-613-616 500 mL Poly Bottle
18-613-617 1 L Poly Bottle
18-613-618 4 L Poly Bottle
18-613-620 20 L Poly Drum
18-613-621 200 L Drum
18-613-619 2.5 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-613-615 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1310100MLPL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5

Specifications

Quantity 100 mL
Percent Purity 48 to 52%
Grade Reagent
Identification To Pass Test
Packaging Poly Bottle
Chemical Name or Material Sodium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.