Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 6.25 N, Ricca Chemical
SDP

Catalog No. 74675 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
74675 20 L Cubitainer
R74670004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74675 Supplier Ricca Chemical Company Supplier No. 74675
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 65.0, 32.76
CAS Max % 68.0, 34.1
Concentration or Composition Notes 6.25Normal
Quantity 20 L
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
Grade Laboratory
Packaging Cubitainer
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.