Learn More
Hach Company Sodium Hydroxide Digital Titrator Cartridge, 8.00 N
Supplier:  Hach Company 1438101
Encompass
For determination of acid concentration by Digital Titrator titration. Digits x 0.01 = meq
Catalog No. 50-199-5012