Learn More
Sodium Hydroxide, ≥98% (Acidimetric), BioXtra, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
- Includes application-specific formulations, such as tablets and prepared solutions.
Specifications
Specifications
| Linear Formula | NaOH |
| UN Number | UN1823 |
| Quantity | 1 kg |
| Formula Weight | 40g/mol |
| Percent Purity | ≥98% (acidimetric) |
| Grade | Certified |
| Vapor Density | >1 (vs air) |
| Vapor Pressure | 18 mmHg (20°C), 3 mmHg (37°C) |
| Chemical Name or Material | Sodium hydroxide |
Safety and Handling
| Hazard Statement | P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |
| RTECSNumber | WB4900000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.