Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium hydroxide ACS reage500G
Catalog No. 502898957
Encompass
Sodium hydroxide (NaOH) also known as caustic soda is a water soluble inorganic base with a wide range of industrial application such as titration disSOLUTIONon testing and in impinger to remove acidic gases. It participates in the oxidation of glycerol catalyzed by Au/charcoal or Au/graphite. It participates in solvent-free aldol condensation reaction.
Catalog No. 50-289-8957 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2214656X500G
Sodium hydroxide (NaOH) also known as caustic soda is a water soluble inorganic base with a wide range of industrial application such as titration disSOLUTIONon testing and in impinger to remove acidic gases. It participates in the oxidation of glycerol catalyzed by Au/charcoal or Au/graphite. It participates in solvent-free aldol condensation reaction.