Learn More
Sodium Hydroxide, Certified, 21.5% (w/w) ±1.0% (w/w), 6.64N (6.64M), LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| Color | Colorless |
| CAS Max % | 78.5, 21.5 |
| Linear Formula | NaOH |
| Concentration | 6.64N (6.64M) |
| UN Number | UN1824 |
| Density | 1.23g/mL |
| Solubility Information | Soluble in water |
| Quantity | 1 L |
| Formula Weight | 40 |
| Percent Purity | 21.5% ±1.0% (w/w) |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves/eye protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.