Learn More
Sodium hydroxide, Concentrate 0.1 M NaOH in Water (0.1 N), Eluent, Concentrate for IC, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Chemical Name or Material | Sodium Hydroxide |
| CAS | 1310-73-2 |
| Linear Formula | NaOH |
| MDL Number | MFCD00003548 |
| Synonym | Sodium hydroxide solution |
| Quantity | 1 L |
| Molecular Weight (g/mol) | 40.00 |
| Formula Weight | 40.00 |
| Grade | Not Available |
| Concentration or Composition (by Analyte or Components) | 0.1 M NaOH aq. soln. (0.1 N) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.