Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide concentrate, For 1 L standard solution, 0.01 M NaOH (0.01 N), Solstice

Catalog No. 60046708 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-708 6 x 1 Ea.
60-026-79 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-708 Supplier Solstice Supplier No. 382276X1EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for 1 L standard solution, 0.01 M NaOH (0.01 N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 100°C
CAS Min % ≥99.0000%, ≥0.5000%
CAS Max % <1.0000%
Linear Formula NaOH
UN Number UN1824
Density 1.000 g/cm3
Quantity 6 x 1 Ea.
Formula Weight 40
Grade Not Available
Packaging Ampoule
Vapor Pressure 3 mmHg (37°C)
Physical Form Concentrate
Chemical Name or Material Sodium hydroxide concentrate, For 1 L standard solution, 0.01 M NaOH (0.01 N)
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P337 + P313
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Sodium hydroxide solution
Recommended Storage Room Temp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.