Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Dilution Solution, 0.16% (w/v), Ricca Chemical
SDP

Catalog No. 747416 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7474-16 500 mL Natural Poly Bottle
7474-32 1 L Natural Poly Bottle
7474-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 7474-16 Supplier Ricca Chemical Company Supplier No. 747416
Add to Cart
Add to Cart

For Dilution of Cyanide Standards

Chemical Identifiers

CAS 1310-73-2

Specifications

Quantity 500 mL
Percent Purity 0.16%
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.16% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Hydroxide Dilution Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.