Learn More
Strem, An Ascensus Company CAS# 12179-02-1. 50g. Sodium hydroxide monohydrate (99.99%-Na) PURATREM. MFCD00149229
SDP
Catalog No. 5090113814
Encompass
CAS# 12179-02-1. 50g. Sodium hydroxide monohydrate (99.99%-Na) PURATREM. MFCD00149229. Molecular Weight: 40.00 (58.01). Molecular Formula: NaOH∙H2O. Color/form: white xtl. Strem# 93-1064. http://www.strem.com/catalog/v/93-1064/
Catalog No. 50-901-13814 Supplier Strem, An Ascensus Company Supplier No. 93106450G
CAS# 12179-02-1. 50g. Sodium hydroxide monohydrate (99.99%-Na) PURATREM. MFCD00149229. Molecular Weight: 40.00 (58.01). Molecular Formula: NaOH∙H2O. Color/form: white xtl. Strem# 93-1064. http://www.strem.com/catalog/v/93-1064/