Learn More
Alkali Scientific Sodium Hydroxide (NaOH) Pellets, ACS Grade, 1 Kg
SDP
Catalog No. 5000660608
Encompass_Preferred
Sodium Hydroxide (NaOH) Pellets (1kg, ACS Grade) is a high-purity reagent used in a variety of laboratory applications that require a strong base. Sodium hydroxide is commonly used for titrations, pH adjustments, and chemical syntheses in analytical chemistry and organic chemistry. It plays a critical role in the preparation of sodium-based buffers, the production of soap in saponification reactions, and the neutralization of acids. The ACS-grade quality ensures the reagent meets high standards of purity, making it suitable for sensitive scientific research and industrial applications where precise pH control is necessary.
Catalog No. 50-006-60608 Supplier Alkali Scientific Supplier No. UMH001CP
Sodium Hydroxide (NaOH) Pellets (1kg, ACS Grade) is a high-purity reagent used in a variety of laboratory applications that require a strong base. Sodium hydroxide is commonly used for titrations, pH adjustments, and chemical syntheses in analytical chemistry and organic chemistry. It plays a critical role in the preparation of sodium-based buffers, the production of soap in saponification reactions, and the neutralization of acids. The ACS-grade quality ensures the reagent meets high standards of purity, making it suitable for sensitive scientific research and industrial applications where precise pH control is necessary.