Learn More
Supply Solutions Avantor Performance Materials - Sodium Hydroxide Pellets. Size: 12kg; Grade: AR (ACS); Assay: 98%.; Size: 12kg; Grade: AR (ACS); Assay: 0.98; CAS: 1310-73-2; Hazardous: Y; Container Material: PE Pail
Catalog No. 50878537
Encompass_Preferred
Avantor Performance Materials - Sodium Hydroxide Pellets. Size: 12kg; Grade: AR (ACS); Assay: 98%.; Size: 12kg; Grade: AR (ACS); Assay: 0.98; CAS: 1310-73-2; Hazardous: Y; Container Material: PE Pail; Formula:NaOH; Formula Weight:40.00; Specific Gravity: 1L = 2.13kg
Catalog No. 50-878-537 Supplier Supply Solutions Supplier No. MACRON 770888
Avantor Performance Materials - Sodium Hydroxide Pellets. Size: 12kg; Grade: AR (ACS); Assay: 98%.; Size: 12kg; Grade: AR (ACS); Assay: 0.98; CAS: 1310-73-2; Hazardous: Y; Container Material: PE Pail; Formula:NaOH; Formula Weight:40.00; Specific Gravity: 1L = 2.13kg