Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide pellets, 99.0%, for analysis EMSURE™, MilliporeSigma™

Catalog No. M1064981000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1064981000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1064981000 Supplier MilliporeSigma Supplier No. 1.06498.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity sodium hydroxide pellets designed for analytical applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym Soda caustic
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH > 14 (100 g/L, H2O, 20°C)
Melting Point 319 to 322°C
Assay Percent Range ≥ 99.0%
Density 2.13 g/cm3 (at 20°C)
Solubility Information 1090 g/L
Quantity 1 kg
Percent Purity ≥99%
Grade Not Available
Packaging Plastic Bottle
Chemical Name or Material Sodium Hydroxide
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.