Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Pellets ACS/USP/NF/FCC, Greenfield

Catalog No. 502946049
Encompass
Change view
Click to view available options
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6049 Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6049 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P2890NFFCCDM50
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2
Molecular Weight (g/mol) 40

Specifications

Quantity 50 kg
Percent Purity > 97%
Packaging Fiber Drum
Chemical Name or Material Sodium Hydroxide Pellets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.