Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, Pellets, BiotechGrade, 95-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606571 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-571 4 x 2.5 kg Poly Bottle
18-606-568 500 g Poly Bottle
18-606-570 2.5 kg Poly Bottle
18-606-569 6 x 500 g Poly Bottle
18-606-572 12 kg Poly Pail
18-606-573 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-606-571 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S13012.5KGCS4
Only null left
Add to Cart
Add to Cart

S1301, 1310-73-2, NaOH

Spectrum™ Chemical Sodium Hydroxide, Pellets, BiotechGrade or caustic soda, is an inorganic, highly basic compound that is water soluble, as well as soluble in ethanol and methanol. One of its many characteristics is that it's a deliquescent alkali and absorbs carbon dioxide and moisture from the air. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

Color White
CAS Max % 1
Quantity 4 x 2.5 kg
Percent Purity 95 to 100.5%
Grade Biotech
Odor Odorless
Packaging Poly Bottle
Physical Form Pellet
Chemical Name or Material Sodium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.