Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, Pellets (Max. 0.02% K), For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1064699012 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
12 kg
Packaging:
Plastic Bottle
Plastic Bucket
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1064699012 12 kg Plastic Bucket
M1064691000 1 kg Plastic Bottle
M1064695000 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1064699012 Supplier MilliporeSigma Supplier No. 1.06469.9012
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH <14 (100g/L, H2O, 20°C)
Color White
Melting Point 319°C to 322°C
Density 2.13 g/3 (20 °C)
Solubility Information 1090g/L
Quantity 12 kg
Formula Weight 40
Percent Purity 99 to 100%
Grade ACS
Packaging Plastic Bucket
Physical Form Solid
Chemical Name or Material Sodium Hydroxide
Show More Show Less
EINECSNumber 215-185-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.