Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, Puriss., meets analytical spec. of BP, NF, Ph. Eur., E524, 98-100.5%, pellets, Solstice

Catalog No. 6001477 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-014-77 1 kg
60-014-78 5 kg
60-044-580 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-014-77 Supplier Solstice Supplier No. 062031KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, NF, Ph. Eur., E524, 98-100.5%, pellets

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number 1823
Solubility Information Soluble in water, 1260g/L at 20°C
Quantity 1 kg
Formula Weight 40g/mol
Percent Purity 98 to 100.5%
Grade puriss.
Vapor Density >1 (vs air)
Vapor Pressure 3mmHg (37°C), 18mmHg (20°C)
Chemical Name or Material Sodium Hydroxide, Meets analytical specification of Ph. Eur.BP, NF
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H314
EINECSNumber 215-185-5
RTECSNumber WB4900000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.