Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., K ≤0.02%, ≥98%, pellets, Honeywell Fluka™

Catalog No. 60045835 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-835 4 x 5 kg
60-014-25 5 kg
60-045-836 6 x 1 kg
60-014-24 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-045-835 Supplier Honeywell Chemicals Supplier No. 306204X5KG
Only null left
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., K ≤0.02%,≥98%, pellets

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 1,390°C
Melting Point 318°C (literature)
CAS Max % 100.0000%
Linear Formula NaOH
UN Number UN1823
Density 2.13 g/cm3
Quantity 4 x 5 kg
Formula Weight 40.00
Percent Purity ≥98%
Grade ACS Reagent/Reagent Ph. Eur.
Packaging Poly Bottle
Physical Form Pellet
Chemical Name or Material Sodium hydroxide, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sodium hydroxide, solid
EINECSNumber 215-185-5
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.