Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium hydroxide puriss. p500G
Catalog No. 502897298
Encompass
Sodium hydroxide (NaOH) is an inorganic base. It participates in the oxidation of glycerol catalyzed by Au/charcoal or Au/graphite. It also participates in solvent-free aldol condensation reaction.
Catalog No. 50-289-7298 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 71690500G
Sodium hydroxide (NaOH) is an inorganic base. It participates in the oxidation of glycerol catalyzed by Au/charcoal or Au/graphite. It also participates in solvent-free aldol condensation reaction.