Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, Reagent Grade, 97%, 20 to 40 Mesh, Solstice

Catalog No. 60046650 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Drum with Inliner
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-650 2.5 kg Poly Bottle
60-046-649 12 kg Drum with Inliner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-650 Supplier Solstice Supplier No. 3671762.5KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade, beads, 20-40 mesh, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless or White
Boiling Point 1390°C
Melting Point 318°C
Linear Formula NaOH
UN Number UN1823
Density 2.13g/cm3
Quantity 2.5 kg
Formula Weight 40g/mol
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Vapor Density >1 (vs air)
Vapor Pressure 18mmHg (20°C), 3mmHg (37°C)
Physical Form Beads
Chemical Name or Material Sodium hydroxide
Show More Show Less
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 8/PGII
RTECSNumber 215-185-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.