Learn More
Andwin Scientific Sodium Hydroxide Solution 6.0N, 1310-73-2, MFCD00003548, 1L
SDP
Catalog No. NC1395393
Encompass_Preferred
Formula: NaOH, MW: 40 g/mol, Boiling Pt: 109 °C (1013 hPa), Melting Pt: 323 °C
Catalog No. NC1395393 Supplier Andwin Scientific Supplier No. BDH72261
Formula: NaOH, MW: 40 g/mol, Boiling Pt: 109 °C (1013 hPa), Melting Pt: 323 °C