Learn More
Hach Company Sodium Hydroxide Standard Solution, 10 N, 500 mL
Supplier:  Hach Company 2545049
Encompass
For sample pH adjustments in ammonia determenations by ion selective electrode.
Catalog No. 50-199-6202