Learn More
Description
- Sodium hydroxide solution is used as a key reactive medium or treatment agent across diverse applications in environmental science, materials engineering, and chemical synthesis.
- In both sets of studies, sodium hydroxide solution enhances material performance, such as improving adsorption capacity, strengthening geopolymers and stabilized soils, or optimizing functional properties of processed materials.
- Research in both groups employs sodium hydroxide solution in aqueous systems to modify or process solid matrices, including biochar, soils, polymers, and biomaterials.
- Across all referenced work, sodium hydroxide solution plays a critical role in developing advanced technologies and processes that support industrial, environmental, and biotechnological applications.
Specifications
Specifications
| Chemical Name or Material | Sodium Hydroxide Solution |
| CAS | 1310-73-2, 7732-18-5 |
| Concentration | 0.1 mol/L (0.1 N) |
| Quantity | 1 Bottle |
| Molecular Weight (g/mol) | 40.00 |
| Percent Purity | Unknown |
| Packaging | Plastic Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.