Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide solution, Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N), Solstice

Catalog No. 6002632 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Packaging:
Plastic Bottle
VOLPAC™ CONTAINER
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-026-32 5 L VOLPAC™ CONTAINER
60-026-30 1 L Plastic Bottle
60-026-31 5 L Plastic Bottle
60-026-29 10 L VOLPAC™ CONTAINER
60-046-557 6 x 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-026-32 Supplier Solstice Supplier No. 352635LVP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, Reag. Ph. Eur., 0.1 M NaOH (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
Concentration 0.1M
UN Number NONH for all modes of transport
Quantity 5 L
Formula Weight 40g/mol
Packaging VOLPAC™ CONTAINER
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.