Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium hydroxide solution, 1310-73-2, MFCD00003548, 100mL
Catalog No. 501656892
Encompass
Linear Formula NaOH, Molecular Weight 40.00, 1.0 N, BioReagent, suitable for cell culture
Catalog No. 50-165-6892 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S2770100ML
Linear Formula NaOH, Molecular Weight 40.00, 1.0 N, BioReagent, suitable for cell culture