Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide solution, 30.0%, Suprapur™, MilliporeSigma™

Catalog No. M1055892500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1055892500 2.5 L
M1055890250 250 mL
M1055891000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1055892500 Supplier MilliporeSigma Supplier No. 1055892500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14 (H2O, 20°C)
Assay Percent Range ≥ 30.0%
Density 1.33 g/cm3 (at 20°C)
Quantity 2.5 L
Formula Weight 40.00
Percent Purity 30%
Grade Reagent
Packaging Plastic Bottle
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Recommended Storage Store at 15 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.