Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Solution, 50% (w/w), MilliporeSigma™

Catalog No. MSX05973 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX05973 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX05973 Supplier MilliporeSigma Supplier No. SX05973

missing translation for 'validMainframeMsg'

For General Chemistry

Recommended Storage

Store at room temperature

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Melting Point 12°C
Boiling Point 135°C (1013hPa)

Specifications

Packaging Plastic Bottle
Vapor Pressure 2 hPa (20 °C)
pH 14 (89 g/L, H2O, 20°C)
Quantity 500 mL
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.