Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Solution, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1055912500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1055912500 2.5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1055912500 Supplier MilliporeSigma Supplier No. 1055912500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium hydroxide solution for analysis, specifically formulated for the determination of nitrogen.

  • Used for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14 (at 20°C)
Color Colorless
UN Number UN 1824
Quantity 2.5 L
Percent Purity >27.0%
Grade Analytical
Packaging Plastic bottle
Physical Form Solution
Chemical Name or Material Sodium hydroxide solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.