Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Hydroxide Solutions, 1M/1N
SCI_ED

Catalog No. S25549A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
1M Solution
1N Solution
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25549A 1 L 1M Solution
S25549 1 L 1N Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25549A

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Linear Formula NaOH
Quantity 1 L
Formula Weight 4
Packaging 1M Solution
Chemical Name or Material Sodium Hydroxide Solutions, 1M/1N
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 8, II, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.