Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide-Sulfide, Ricca Chemical
SDP

Catalog No. 74801 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7480-1 4 L Natural Poly Bottle
74805 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7480-1 Supplier Ricca Chemical Company Supplier No. 74801
Add to Cart
Add to Cart

For Analysis of Nitrogen (Kjeldahl) Using Mercrury Catalyst

Chemical Identifiers

Molecular Formula HNaO
Molecular Weight (g/mol) Mixture
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Min % 0.74, 18.53
Color Amber
CAS Max % 0.77, 19.29
CAS 10294-66-3, 1310-73-2, 7732-18-5, 37488-75-8
Physical Form Liquid
Quantity 4 L
Packaging Natural Poly Bottle
Grade Laboratory
Chemical Name or Material Sodium Hydroxide-Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.